Puritan® Small Rectangular Foam Tipped Applicator, Polypropylene Handle, Sterile, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
This swab’s small rectangular foam tip has a high collection capacity ideal for diagnostics, forensics, and buccal swabbing as well as industrial cleaning
Made in the USA.
The tip is attached to a rigid polypropylene handle
Certifications: CE marked.
Ordering information: Packaged as 10 boxes of 50 individually wrapped swabs.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together