Puritan® Small Tapered Foam Tipped Applicator, Polystyrene Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
Certificates
About this item
Swabs are small to ensure patient comfort
Made in the USA.
Feature a molded polystyrene handle and are ideal for specimen collection and ophthalmology applications. Swab with reclosable dry transport tube is ideal for transporting specimens for lab work.
Certifications: CE marked.
Ordering information: Packaged as 10 boxes of 50 individually wrapped swabs.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together