Puritan® Calgiswab® Mini Calcium Alginate Tipped Applicator, Aluminum Handle, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
This specialty applicator is tipped with calcium alginate fiber, which can be inhibitory to many strains of bacteria
It is ideal for specimen collection for testing and screening, as well as small cavities and wound care. It has a flexible aluminum handle and comes individually wrapped
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together