VWR® Destaining Bags
97062-968
: VWR
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Size:25 Bags
- Cat. no.:97062-968
- Supplier No.:E732-25
Bags safely remove ethidium bromide from solution during overnight treatment
Each bag extracts up to 5mg of ethidium bromide, minimizing personnel exposure to toxic material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration.