VWR® TRIS-Borate-EDTA Buffer, 10X Liquid Concentrate, Ultra Pure Grade
: VWR
TBE is optimized for DNA electrophoresis techniques using both acrylamide and agarose.A concentrated (10X), pH adjusted solution of Ultra Pure Grade Tris Buffer, ACS Grade Boric Acid & EDTA which when reconstituted to a working (1X) Solution concentration, contains 0.089M Tris, 0.089M Borate & 0.002M EDTA.
- High Buffering Capacity
- High Ionic Strength
- Permits Extended Electrophoresis Runs with no Buffer Recirculation
- Convenient Packaging Options Minimize Weighing & Mixing
*Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.
No DNase, Protease, RNase. pH (1:10) @25°C is 8.2 - 8.4. Conductivity (1:10) @25°C is 750-1250 umhos.
Frequently Bought Together