VWR® PCR DNA Marker
Catalog # 97063-054
Supplier: VWR
CAS Number:
Specifications
- Description:PCR DNA Marker™
- Size:100 rxn
- Cat. no.:97063-054
- Application:Sufficient for 100 lanes
- Supplier No.:E854-100RXN
Specifications
About this item
8 fragments ranging from 50 to 2000 bp for easy band identification
High-resolution marker optimal for size estimation of PCR fragments. Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.