VWR® 2XYT Medium Broth
97063-442
: VWR
:
- Description:2XYT MEDIUM BROTH
- Size:500 g
- Cat. no.:97063-442
- Supplier No.:J902-500G
2XYT Medium Broth powder is used to prepare rich broth for the propagation of M13 bacteriophage.
- Save time using premixed and ready-to-use media
- Homogenous and high quality powders
- Light beige, free flowing powder
31g of powder when dissolved per liter of deionized water will yield formulation- 16 grams/L of cesein peptone, 10 grams/L of casamino acids and 5 grams/L of sodium chloride.