VWR® Yeast Nitrogen Base (YNB)
Supplier:   VWR
VWR® Yeast Nitrogen Base (YNB)
Supplier:  VWR
Certificates
VWR® Yeast Nitrogen Base (YNB)
VWR® Yeast Nitrogen Base (YNB)
2 Options Available Below
Product Details & Documents
YNB is a base medium for preparation of minimal and synthetic defined yeast media.
  • Animal origin
  • Suitable for studying strains of yeast requiring certain vitamins
  • Base medium
  • Prepares minimal and synthetic defined yeast
YNB is a base medium for preparation of minimal and synthetic defined yeast media. It is a suitable medium for studying strains of yeast that require certain vitamins.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Utilisation
    Availability
    Price
  • YNB Without Amino Acids
    100 g
    Used for yeast classification based on amino acid and carbohydrate requirements.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100g)
    Each (100g) - $246.64
    $246.64
    Log in pour la disponibilité
    Case of 12 (100g)
    Case of 12 (100g) - $2,959.36
    $2,959.36
    Each (100g)
       $246.64
    Case of 12 (100g)
       $2,959.36
    Supplier Number: J386-100G
  • YNB Without Amino Acids
    500 g
    Used for yeast classification based on amino acid and carbohydrate requirements.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500g)
    Each (500g) - $1,135.34
    $1,135.34
    Log in pour la disponibilité
    Case of 12 (500g)
    Case of 12 (500g) - $13,624.08
    $13,624.08
    Each (500g)
       $1,135.34
    Case of 12 (500g)
       $13,624.08
    Supplier Number: J386-500G
  • YNB Without Amino Acids or Ammonium Sulfate
    100 g
    Used for yeast classification based on carbon and nitrogen requirements.
    Discontinued
    Supplier Number: -