VWR® Malt Extract
: VWR
VWR® offers high purity extracts used for media supplements.
- Used for the preparation of media for the detection of yeasts and molds. Hazards, N/A. Bacteriological Grade
VWR® offers high purity extracts used for media supplements.
- Used for the preparation of media for the detection of yeasts and molds. Hazards, N/A. Bacteriological Grade
- Catalog No:
- 97063-426
- 97063-428
- 97063-430
- Total nitrogen:
- 2.0%
- 2.0%
- 2.0%
- Coliforms:
- 10/g
- 10/g
- 10/g
- Ash content:
- 5%
- 5%
- 5%
- Loss on drying:
- 5.0%
- 5.0%
- 5.0%
- pH:
- 6% Water, 4.5 to 5.7 (at 25 °C)
- 6% Water, 4.5 to 5.7 (at 25 °C)
- 6% Water, 4.5 to 5.7 (at 25 °C)
- Solubility:
- 6%, Water, Pass
- 6%, Water, Pass
- 6%, Water, Pass
- Storage conditions:
- Cold
- Cold
- Cold
Product Family Options
- Taille
- -: -
Specifications
Cat. No.97063-426Total nitrogen2.0%Coliforms10/gAsh content5%Loss on drying5.0%pH6% Water, 4.5 to 5.7 (at 25 °C)Solubility6%, Water, PassStorage conditionsCold - -: -
Specifications
Cat. No.97063-428Total nitrogen2.0%Coliforms10/gAsh content5%Loss on drying5.0%pH6% Water, 4.5 to 5.7 (at 25 °C)Solubility6%, Water, PassStorage conditionsCold - 500 gEach (500g)$181.45Case of 4 (500g)$617.95: J873-500G
Specifications
Cat. No.97063-430Total nitrogen2.0%Coliforms10/gAsh content5%Loss on drying5.0%pH6% Water, 4.5 to 5.7 (at 25 °C)Solubility6%, Water, PassStorage conditionsCold