ContactUsLinkComponent
VWR® TRIS Glycine Buffer
  97063-782
 :  VWR
 :  
undefined
VWR® TRIS Glycine Buffer
  97063-782
 :  VWR
 :  M115-40L
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    TRIS-GLYCINE BUFFER, 10X READY-PACK™
  • Size:
    40 L
  • Cat. no.:
    97063-782
  • Application:
    Enough powder to make 40 L of 1X buffer. Each pack makes 1L of 10X concentrate. Add methanol to make standard transfer buffer.
  • Supplier No.:
    M115-40L
Following electrophoresis, proteins are frequently transferred from the polyacrylamide gel onto PVDF or nitrocellulose membranes for use in immunoblotting or sequencing
  • Convenience of liquid concentrates or powder blend
  • Consistent performance and quality
Although there are different methods by which to achieve transfer, the one most routinely used is electroblotting. Electroblotting utilizes conductive buffers that do not interfere with proteins binding to the membrane and can be performed in a vertical buffer tank or in a semi-dry blotting apparatus.