Rapidblock™ Blocking Solution, 10X, AMRESCO
 :   VWR
Rapidblock™ Blocking Solution, 10X, AMRESCO
 :  VWR
Rapidblock™ Blocking Solution, 10X, AMRESCO
Rapidblock™ Blocking Solution, 10X, AMRESCO
Solution reduces blocking time to 5 minutes for Western and dot blotting procedures on PVDF and nitrocellulose membranes.
  • Only 5 Minutes for PVDF or Nitrocellulose Blocking
  • Reduce Background and Cross-Reactivity
  • Enhance Chemiluminescent Signals
  • Protein-Free Formulation
This protein-free formulation minimizes cross-reactivity and non-specific antibody binding, generating blots with low background and enhanced signal-to-noise ratio. Results meet or exceed those obtained with buffers containing dried milk or BSA, which both require 1 hour of blocking time.
Product Family Options