VWR® Stripping Buffers
Fournisseur: VWR
Certificats
À propos de cet article
VWR® offers three stripping buffers to accommodate the wide range of conditions that may be needed to remove different antibodies from blots.
- ReView™ Buffer Solution
- Save money and time
- Compare detection of different antibodies on a single blot
- Gentle ReView™ Buffer
- Odor-free, room-temperature procedure—No hood needed
- Minimize loss of valuable antigen
- Vantage ReView™ Stripping B
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Environmentally Preferable:Safer Human and Environmental Health