VWR® Drosophila Vial Plugs, Cellulose Acetate
Bonded dense-weave cellulose acetate plugs provide a better barrier to mites with minimal shedding.
- Cellulose acetate fabrication
- Excellent barrier to mites
- Minimal shedding
Reduce evaporation to keep food fresh longer.
Frequently Bought Together