HBIO BTXpress Solution, BTX
HBIO BTXpress Solution, BTX
Supplier:  Harvard Apparatus
Certificates
HBIO BTXpress Solution, BTX
HBIO BTXpress Solution, BTX
7 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered 'hard to transfect' by chemical and other non-viral methods.
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types
BTXpress Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you.
As a universal solution, the BTXpress Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXpress Electroporation Solution is offered as a kit including the BTX Cuvettes Plus with transfer pipettes or as a buffer alone.
 : Trial Sample Kit includes 2 ml volume solution with five 2 mm gap cuvettes and five 4 mm gap cuvettes. The 2 ml volume is the solution only and it requires cuvettes. The The 5 ml volume is the solution only for up to 50 reactions. It requires twenty 4 mm gap cuvettes or fifty 2 mm gap cuvettes. The 10 ml volume is the solution only for up to 100 reactions. It requires forty 4 mm gap cuvettes or one hundred 2 mm gap cuvettes. The 20 Reaction Kit includes 5 ml volume solution and twenty 4 mm gap cuvettes. The 40 Reaction Kit includes 5 ml volume solution and forty 4 mm gap cuvettes. The 100 Reaction Kit includes 5 ml volume solution and one hundred 4 mm gap cuvettes.
Product Family Options
BTXpress Solution
Informations sur le produit
  • Désignation
    Taille
    No. of Reactions
    Availability
    Price
  • BTXpress Solution Only, 2 mL
    2.0 ml
    20
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $396.78
    $396.78
    Prix promotionnel pour les nouveaux clients
    Each
       $396.78
    Supplier Number: 45-0801
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • BTXpress™ Solution, 5 mL
    5.0 ml
    50
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,052.09
    $1,052.09
    Each
       $1,052.09
    Supplier Number: 45-0802
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • BTXpress™ Solution, 10 mL
    10.0 ml
    100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,502.44
    $1,502.44
    Each
       $1,502.44
    Supplier Number: 45-0805
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
BTXpress Solution Kit
Informations sur le produit
  • Désignation
    Taille
    No. of Reactions
    Availability
    Price
  • BTXpress solution kit
    20 in 4 mm gap cuvettes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,054.56
    $1,054.56
    Each
       $1,054.56
    Supplier Number: 45-0804
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • BTXpress solution kit
    40 in 4 mm gap cuvettes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,665.47
    $1,665.47
    Each
       $1,665.47
    Supplier Number: 45-0807
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • BTXpress solution kit
    50 in 2 mm gap cuvettes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,887.71
    $1,887.71
    Each
       $1,887.71
    Supplier Number: 45-0803
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • BTXpress solution kit
    100 in 2 mm gap cuvettes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,566.16
    $2,566.16
    Each
       $2,566.16
    Supplier Number: 45-0806
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.