Duck® Removable Poster Putty®, Essendant
Adhesive mounting putty, 2 oz.
Catalog # 500000-051
Supplier: Janitorial Supplies
Supplier #: DUCPTY2
Jump to:
Product Details & Documents
Great for temporary mounting of paper items such as posters, charts and decorations
- Safe, clean alternative to tape, nails, glues, tacks and staples
- Non-staining on most nonporous surfaces
- Nontoxic
- Removable and reusable again and again
Putty will not dry out and won't damage mounting surface.
Certifications: Conforms to ASTM D-4236
Specifications
- Volume:2 oz.
- Désignation:Adhesive mounting putty
- Cat. No.:500000-051
- Application:Adheres paper to clean dry surfaces