FlexBrush™ Brushes and Covers, Micronova
Supplier: Micronova
Certificates
About this item
A flexible 12" brush with optional covers in polyester or microfiber. The FlexBrush™ with attached handle is threaded and can be used with a STA series plastic, ACME threaded handle for extended reach.
- Ideal for cleaning over and around piping and fittings
- Flexibility of the head allows users to 'shape' the head for custom cleaning applications
- Available in J or Hook shape
- Stainless steel or zinc twisted wire core
- FDA approved materials
Polyester or microfiber covers are lined with 100% knit polyester to easily slide onto the FlexBrush™, containing the bristles. The covers provide the cleaning of a mop with the flexibility of a brush.
Specifications
- Catalog No:
- 68400-718
- 76214-300
- 10153-664
- 89092-120
- 68400-720
- 10126-342
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing