Accessories
Clamps for 35/20 spherical joints
Catalog # 60004-978
Supplier: Kemtech America
Fournisseur #: CC3525
Jump to:
Product Details & Documents
Kemtech pinch clamp designed for spherical joints.
- Stainless steel construction
- Adjustable metal tensioner
For use with 35/20, 35/25 ball joint, or #25 O-ring joints.
Spécifications
- Désignation:Clamps for 35/20 spherical joints
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- PTFE plug, 4 mm
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 2 mm PTFE Plug