Stage Micrometer Slide, Leica Microsystems
Catalog # CA16002-166
Supplier: Leica Microsystems
Specifications
- Description:Stage micrometer slide for microscope calibrations, 1", 0.001" divisions
- Cat. no.:CA16002-166
- Supplier No.:10447037
Specifications
About this item
For microscope calibrations.
- 1"
- 0.001" divisions