WHEATON® MicroLiter Screw-Thread Headspace Vials, 16 mm, DWK Life Sciences
WHEATON® MicroLiter Screw-Thread Headspace Vials, 16 mm, DWK Life Sciences
WHEATON® MicroLiter Screw-Thread Headspace Vials, 16 mm, DWK Life Sciences
WHEATON® MicroLiter Screw-Thread Headspace Vials, 16 mm, DWK Life Sciences
WHEATON® MicroLiter Screw-Thread Headspace Vials, 16 mm, DWK Life Sciences
These headspace vials offer excellent performance.
  • Caps are made of blue polypropylene
  • Cleaner vials improve performance by eliminating almost all particulates
Vials are made from Type 1 borosilicate glass and work well on PerkinElmer®, Agilent™, CTC Analytics, Shimadzu™, Tekmar®, and other headspace autosamplers. They meet strict tolerances set by the manufacturer of the instrument.
Every glass vial is precleanedl and packaged in an ISO Class 7 (Class 10000) cleanroom, in a box bearing the Class 10K seal. The result is a vial that exceeds most OEM requirements at no additional cost.
Documents
Product Family Options
Vials
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
  • Vial
    10.00 ml
    22×46 mm
    Clear
    Non
    Pack of 100
    Pack of 100 - $115.90
    $115.90
    Pack of 100
       $115.90
     : 16-1000
  • Vial
    20.00 ml
    22×75 mm
    Clear
    Non
    Pack of 100
    Pack of 100 - $117.54
    $117.54
    Pack of 100
       $117.54
     : 16-2000
  • Vial, Silanized
    20.00 ml
    22×75 mm
    Clear
    Non
    Pack of 100
    Pack of 100 - $286.90
    $286.90
    Pack of 100
       $286.90
     : 16-2000-S
  • Vial
    10.00 ml
    22×46 mm
    Amber
    Non
    Pack of 100
    Pack of 100 - $189.12
    $189.12
    Pack of 100
       $189.12
     : 16-6100
  • Vial
    20.00 ml
    22×75 mm
    Amber
    Non
    Pack of 100
    Pack of 100 - $209.00
    $209.00
    Pack of 100
       $209.00
     : 16-6000
Caps
  •  
    Désignation
    Cap Color
    Ø
    Épaisseur
    Septum
  • Headspace ScrewTin™ Screw Cap
    Black
    16 mm
    1.5 mm (0.060")
    PTFE/Butyl
    Pack of 100
    Pack of 100 - $186.20
    $186.20
    Pack of 100
       $186.20
  •  
    Désignation
    Cap Color
    Ø
    Épaisseur
    Septum
  • Headspace ScrewTin™ Screw Cap
    Silver
    16 mm
    1.5 mm (0.060")
    White PTFE/Blue Silicone
    Pack of 100
    Pack of 100 - $406.60
    $406.60
    Pack of 100
       $406.60
  •  
    Désignation
    Cap Color
    Ø
    Épaisseur
    Septum
  • Headspace ScrewTin™ Screw Cap
    Silver
    16 mm
    1.4 mm (0.055")
    Viton
    Pack of 100
    Pack of 100 - $348.48
    $348.48
    Pack of 100
       $348.48
Septa
  •  
    Septum
    Désignation
  • PTFE/butyl
    Septa
    Pack of 100
    Pack of 100 - $108.30
    $108.30
    Pack of 100
       $108.30