VWR® Sterile 70% Isopropanol
Supplier: VWR International
Certificates
About this item
Filtered to 0.2 µm and induction sealed to assure sterility.
- Contains 70% USP grade isopropanol and 30% USP grade deionized water
- Double-bagged and gamma-irradiated to a Sterility Assurance Level of 10-6
- Ideal for use with sterile cleanroom wipers
Excellent for wipedown of isolators, cabinets, laminar flow hoods.
Each bottle is lot coded and has an expiration date.
Suitable for wipedown of products and equipment for pass through to controlled environments. Can be used on gloved hands in sterile environments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together