Puritan® PurCollect Environmental Large Foam Tipped Sampling Swab, Polypropylene Handle with Thumb Stop, Sterile, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
Environmental sampling device features a polypropylene shaft with prominent thumb stop to indicate hand placement
Made in the USA.
Shaft is scored 40mm (19/16") from end of tip to allow easy detachment. Large macrofoam tip for maximum surface collection.
Certifications: CE marked.
Ordering information: Packaged as 10 boxes of 50 individually wrapped swabs.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together