Puritan® DNA-Free Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
For crime scene investigation or specimen collection
Sturdy wood shaft. Absorbent cotton tip.
Certifications: Certified DNA-free and RNase-free.
Ordering information: Packaged 100 individually wrapped applicators per box, 10 boxes per case.
Frequently Bought Together