PurSwab® Mini-Tip Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
For removing epoxy residue from connectors and general cleaning
Made in the USA.
Black polypropylene shaft. Flexible mini foam tip. Can be used with IPA and DI water or dry. Lint and residue free. Nonsterile.
: CE marked.
: Packaged as 2 bags of 500 swabs per bag.
Frequently Bought Together