Puritan® Thin Paddle Shaped Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
Cleaning swab is used in manufacturing, or refurbishing of electronic and communication devices as well as removing residues and particulates
Polypropylene shaft. Flexible thin paddle tip. Low in NVR’s and particulates. Chemical resistant. Lint free. Nonsterile. Made in the USA.
Certifications: CE marked.
Ordering information: Packaged 100 per pack, 10 packs per case.
Frequently Bought Together