AlphaSat® Vectra® Alpha® 10 Wiper, Texwipe®
Catalog # CATWTX8410
Supplier: ITW Texwipe
Specifications
- Cat. no.:CATWTX8410
- W×L:22.9 x 22.9 cm (9 x 9")
- Supplier No.:TX8410
Specifications
About this item
Ultraclean wipers presaturated with 100% semiconductor-grade isopropyl alcohol.
Wipers comply with FDA and RoHS requirements. Bag-to-bag traceability.
Wipers are Vectra-processed, 100% continuous-filament double-knit polyester with thermally sealed borders. Superior strength guards against abrasion under rigorous use. Excellent for cleaning surfaces sensitive to water contamination, such as vacuum process chambers for etching and chemical vapor deposition (CVD) equipment. Suitable for maintenance of manufacturing equipment for semiconductor, disk media, and flat panels. Ideal for cleaning activities where use of flammable liquids is restricted or for environments with stringent VOC regulations.
Ordering information: Double-bagged and packaged in a red, easy-to-use reclosable slider bag with internal security seal.