Ethyl Vinyl Acetate (EVA) Bag Systems, Sterile, Advanced Scientifics
Fournisseur: Advanced Scientific, Inc.
Certificats
À propos de cet article
Ethyl vinyl acetate (EVA) single-use bag systems are produced from a Four-layer 12.5 mil coextrusion film for a durable barrier.
- Irradiated and sterile
- Eliminating the concern of cross-contamination
Manufactured in an ISO Class 7 (FED-STF-209E Class 10,000/M5.5) cleanroom environment. Ideal for biopharmaceutical, laboratory, and diagnostic applications.
Certifications: EVA film meets United States Pharmacopoeia Class VI Plastics Guidelines. ISO 13485 certified. Complies with cGMP manufacturing regulations. FDA registered.
Frequently Bought Together