DuPont™ Tyvek® IsoClean® Frocks with High Mandarin Collar and Set Sleeves
: DuPont
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Serged seams
- High mandarin collar with adjustable snaps
- Set sleeve design
- Elastic wrists
- Front snap closure
- Generous sizing and cut
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic openings at wrists and high mandarin collar with adjustable snaps allow for a secure fit. Frocks provide body-length jacket design with front snap closures for easy donning and doffing.
: Bulk packaged garments are bulk packaged in double transparent poly liners.
- Catalog No:
- CA89127-296
- CA89127-300
- CA89127-298
- CA89127-304
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
Frequently Bought Together