CONSTIX®, Sealed Foam Swabs, SF-5 Series, Contec®
Catalogue # 68310-236
Fournisseur: Contec
Small Business Enterprise
Spécifications
- Description:Sealed Foam Swabs, SF-5 Series
- Cat. no.:68310-236
- Handle Length:147.8 mm (5.82")
- Overall Length:163 mm (6.4")
- Head Width:6.4 mm (0.25")
- Head Length:15.2 mm (0.6")
- Handle Width:3 mm (0.12")
- Supplier No.:SF-5
Spécifications
À propos de cet article
CONSTIX® SF-5 swabs consist of a polyurethane foam head bonded to a flex-tip polypropylene handle without the use of adhesives or binders.
- Flexible swab head tip provides good surface contact and controlled pressure to surfaces
The polyurethane foam head provides good particle entrapment and removal. The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. The SF-5 swab is excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-5 is not recommended for use with ketones such as acetone or MEK.
Ordering information: Laundered and double bagged.