CONSTIX®, Cotton Swabs, SC-3 Series, Contec®
Catalog # 68310-334
Supplier: Contec
Small Business Enterprise
Specifications
- Description:Cotton Swabs, SC-3 Series
- Cat. no.:68310-334
- Handle Length:62.5 mm (2.46")
- Overall Length:76 mm (3")
- Head Width:2.6 mm (0.10")
- Head Length:13.5 mm (0.53")
- Handle Width:1.5 mm (0.06")
- Supplier No.:SC-3
Specifications
About this item
CONSTIX® SC-3 double-ended round cotton swabs consist of high-quality, long staple-length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles. The heads are held to the handles using a methyl-cellulose binder.
- Absorbent cotton head is ideal for use with aqueous based solutions
- Tightly wrapped head help maintain low levels of extractable residue
The high-quality cotton fibers provide excellent particle entrapment and retention, as well as exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-cellulose binder which minimizes fiber shedding. The SC series cotton swabs are absorbent to most common solvents including isopropyl alcohol, acetone, and aqueous solutions.