Sampling System, Sterile, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
Jump to:
À propos de cet article
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
6 Options Available Below
Product Details & Documents
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Product Family Options
Informations sur le produit
- CapacitéAvailabilityPrice
- Supplier Number: 4MP0034
- Supplier Number: 4MP0035
- Supplier Number: 4MP0036
- Supplier Number: 4MP0037
- Supplier Number: 4MP0038
- Supplier Number: 4MP0039