DECON-HAND® Cleaning Agent, Veltek Associates
Catalogue # 75795-614
Fournisseur: Veltek Associates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Capacity:16 oz.
- Description:Wall mounted dispenser bottle
- Cat. no.:75795-614
- Supplier No.:DH-100
Spécifications
À propos de cet article
This gelled alcohol hand sanitizer is designed to decrease bacteria on the skin before glove donning.
Requires no water or towels. Sterile product is gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.
The 32 oz bottle is made to custom fit into our ASEPTI-CLEANSE dispensing system that delivers a measured amount of DECON-HAND every time.