VWR® Flat Poly Bags on Rolls
Fournisseur: VWR International
Certificats
À propos de cet article
Flat bags are made with an extra-strong bottom seal or a durable side weld.
- Three times the impact and twice the tensile strength of traditional LDPE bags
- Made with 100% renewable energy
Outstanding vapor and moisture barrier. Ideal for product presentation. Wound on 7.6 cm (3") cores. Core plugs have 2.5 cm (1") Ø holes. Engineered to provide over three times the impact and twice the tensile strength of traditional low-density polyethylene bags.
Certifications: Meet FDA and USDA specifications.
Spécifications
- Catalog No:
- 89072-306
- 89072-308
- 89072-310
- 89072-302
- 89072-304
- Thickness:
- 4 mil
- 4 mil
- 4 mil
- 4 mil
- 4 mil