Puritan® DNA-Free Cotton Tipped Applicator with Transport Tube, Wood Handle, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
For use in specimen collection, wound care, application of topical dressings, cosmetic applications/tattoos, or general purpose
Transport tube packed swabs are certified DNA-free and RNase-free. All swabs are supplied sterile.
Semi-flexible wood shaft. Regular tip.
Frequently Bought Together