Puritan® Tapered Mini Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
This absorbent cotton tipped applicator is produced with high quality USP grade cotton, which is carded and coiled onsite to maintain consistency
It is bonded securely to a sturdy wood handle by an aqueous-based adhesive. The tapered mini cotton tip is ideal for hard-to-reach or small areas. Available nonsterile or sterile.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together