VWR® Polyester Wipers
About this item
Durable wipers resist fiber release and abrasion under rigorous use, even when wiping rough surfaces.
- 100% virgin, interlock knit polyester
- Continuous filament
- No-run material
- Laser cut
- Sealed edges
Laser-cut wipers with sealed edges limit particles and fibers to an absolute minimum, ensuring excellent performance in environments that demand low levels of contaminants.
Made from 100% virgin-grade, continuous-filament polyester in a double-knit, no-run interlock pattern to provide maximum cleanliness and absorbency. Designed for cleaning surfaces and controlling spills in ISO Class 3 (FED-STD-209E Class 1/M1.5) or less critical environments.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers comply with RoHS 2002/95/EC.
Packaging: Packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers are vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with a label on each inner bag.
2 Options Available Below
Product Details & Documents
Durable wipers resist fiber release and abrasion under rigorous use, even when wiping rough surfaces.
- 100% virgin, interlock knit polyester
- Continuous filament
- No-run material
- Laser cut
- Sealed edges
Laser-cut wipers with sealed edges limit particles and fibers to an absolute minimum, ensuring excellent performance in environments that demand low levels of contaminants.
Made from 100% virgin-grade, continuous-filament polyester in a double-knit, no-run interlock pattern to provide maximum cleanliness and absorbency. Designed for cleaning surfaces and controlling spills in ISO Class 3 (FED-STD-209E Class 1/M1.5) or less critical environments.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers comply with RoHS 2002/95/EC.
Packaging: Packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers are vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with a label on each inner bag.
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- 414004-517
- 414004-518
- Basis weight:
- 135± g/m²
- 135± g/m²
- Absorbency - extrinsic:
- 350 ml/m²
- 350 ml/m²
- Absorbency - intrinsic:
- 2.5 ml/g
- 2.5 ml/g
- Absorbency rate:
- <1 sec.
- <1 sec.
- Extractables:
- <0.01 g/m² using deionized water,
<0.02 g/m² using isopropyl alcohol - <0.01 g/m² using deionized water,
<0.02 g/m² using isopropyl alcohol
- Particle generation (>=0,5 µm):
- 4.2 x 10⁶/m²
- 4.2 x 10⁶/m²
- Ions - chloride:
- <0.05 ppm
- <0.05 ppm
- Ions - potassium:
- <0.02 ppm
- <0.02 ppm
- Ions - sodium:
- <0.02 ppm
- <0.02 ppm
Specifications
Product Family Options
Product Information
- ColorPackagingW×LAvailabilityPrice
- White150/Bag22.9x22.9 cm (9x9")Pack of 150$157.51Case of 1200$1,260.08Pallet of 48000$50,403.20Specifications:
More
Specifications
Cat. No.414004-517Basis weight135± g/m²Absorbency - extrinsic350 ml/m²Absorbency - intrinsic2.5 ml/gAbsorbency rate<1 sec.Extractables<0.01 g/m² using deionized water,
<0.02 g/m² using isopropyl alcoholParticle generation (>=0,5 µm)4.2 x 10⁶/m²Ions - chloride<0.05 ppmIons - potassium<0.02 ppmIons - sodium<0.02 ppm - White150/Bag30.5x30.5 cm (12x12")Pack of 150$252.93Case of 900$1,517.58Pallet of 16200$27,316.44Specifications:
More
Specifications
Cat. No.414004-518Basis weight135± g/m²Absorbency - extrinsic350 ml/m²Absorbency - intrinsic2.5 ml/gAbsorbency rate<1 sec.Extractables<0.01 g/m² using deionized water,
<0.02 g/m² using isopropyl alcoholParticle generation (>=0,5 µm)4.2 x 10⁶/m²Ions - chloride<0.05 ppmIons - potassium<0.02 ppmIons - sodium<0.02 ppm