VWR® Laboratory Hygrometer/Thermometer
0...120 °C; 0...100% RH, 1°; 2% RH, 2°C, 4% RH, 127 mm
Catalog # 82021-180
Supplier: VWR International
Jump to:
Product Details & Documents
Durable hygrometer/thermometer is encased in stainless steel and can withstand conditions in incubators, refrigerators, and freezers.
- Stainless steel enclosure
- Durable for withstanding incubators, freezers, and refrigerators
- Easy-to-read dials
A black temperature dial and a red humidity dial are set against a white background. Complete with hole for wall mounting.
Specifications
- Ø:127 mm
- Divisions:1°; 2% RH
- Accuracy:2°C, 4% RH
- Range:0...120 °C; 0...100% RH