Plant Culture Containers, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Designed specifically for plant culture applications, these clear polystyrene containers provide maximum light transmission, promoting fast, strong plant growth.
- Clear polystyrene construction
- Suitable for transporting cultures
- Clear siding promotes strong plant growth
Excellent optical clarity allows for constant viewing of container contents. Containers are also suitable for the safe transport of cultures. Available in complete sets with container and lid, or simply as container without lid.
Frequently Bought Together