Sample Containers with Screw Caps, Polypropylene, BrandTech
Fournisseur: Brandtech
Certificats
À propos de cet article
Translucent polypropylene containers with screw caps
- Chemically resistant polypropylene
- Easy to clean
- Screw on caps for security
Maximum operating temperature: 125 °C (225 °F). Autoclavable.
Frequently Bought Together