VITLAB® Sample Containers with Screw Caps, PFA, BrandTech
À propos de cet article
Containers and screw caps are made of PFA (perfluoroalkoxy copolymer), a fluoropolymer with excellent chemical resistance.
- Trace analysis grade
- Easy to clean
- Screw on caps for security
Surfaces are much less porous than PTFE and are highly cleanable to reduce sample contamination by 'memory effect' from prior samples. Ideal for use in all aspects of ICP-MS and other trace analysis work. Containers provide excellent stability for standards and are manufactured without metal catalysts. Highly translucent for verification of contents. Thermally stable from –200 to 260 °C (–328 to 500 °F). Autoclavable.
4 Options Available Below
Product Details & Documents
Containers and screw caps are made of PFA (perfluoroalkoxy copolymer), a fluoropolymer with excellent chemical resistance.
- Trace analysis grade
- Easy to clean
- Screw on caps for security
Surfaces are much less porous than PTFE and are highly cleanable to reduce sample contamination by 'memory effect' from prior samples. Ideal for use in all aspects of ICP-MS and other trace analysis work. Containers provide excellent stability for standards and are manufactured without metal catalysts. Highly translucent for verification of contents. Thermally stable from –200 to 260 °C (–328 to 500 °F). Autoclavable.
Product Family Options
Informations sur le produit
- CapacitéDésignationDiameter ×HAvailabilityPrice
- 30 ml (1 oz.)With GL40 thread38×54 mmLog in pour la disponibilitéLog in pour la disponibilitéPack of 1Pack of 1 - $280.66$280.66Prix promotionnel pour les nouveaux clientsPack of 1$280.66Supplier Number: 130297
- Supplier Number: 130397
- 90 ml (3 oz.)With GL56 thread54×62 mmLog in pour la disponibilitéLog in pour la disponibilitéPack of 1Pack of 1 - $382.24$382.24Prix promotionnel pour les nouveaux clientsPack of 1$382.24Supplier Number: 130497
- 180 ml (6.1 oz.)With GL56 thread54×112 mmLog in pour la disponibilitéLog in pour la disponibilitéPack of 1Pack of 1 - $462.31$462.31Prix promotionnel pour les nouveaux clientsPack of 1$462.31Supplier Number: 130597