Sample Tubes, 50 mL, Simport Scientific
Sample Tubes, 50 mL, Simport Scientific
Sample Tubes, 50 mL, Simport Scientific
Sample Tubes, 50 mL, Simport Scientific
These polypropylene, conical bottom tubes with self-standing bases have molded-in graduations from 10 to 50 ml in 5 ml increments.
  • Chemical-resistant and shatterproof
  • With a leakproof screw cap, which is particularly important for transporting hazardous material
Ideal for transport and storage of urine, sputum, and most liquids or particulate samples.
Temperature: –90 to 121 °C (–130 to 250 °F).
Product Family Options
  • Capacité
    Ø×L
    Cap Color
    Sterility
  • 50 ml (1.6 oz.)
    30×115 mm (1⅛×4½")
    Yellow
    Non sterile
    Case of 500
    Case of 500 - $384.78
    $384.78
    Case of 500
       $384.78
     : C571-2
  • 50 ml (1.6 oz.)
    30×115 mm (1⅛×4½")
    Green
    Sterile
    Pack of 25
    Pack of 25 - $47.50
    $47.50
    Case of 500
    Case of 500 - $950.00
    $950.00
    Pack of 25
       $47.50
    Case of 500
       $950.00
     : C571-1