Sieve Receiver Pans, W.S. Tyler™
Sieve Receiver Pans are designed for use with 20.3cm (8") or 30.5cm (12") diameter sieves.
Pans with extended rims may be placed within a stack of sieves when sieving two or more samples simultaneously.
Frequently Bought Together