Pre-Cleaned Cubitainer® Containers | Avantor
Pre-Cleaned Cubitainer® Containers
Pre-Cleaned Cubitainer® Containers
Certificates
Pre-Cleaned Cubitainer® Containers
5 Options Available Below
Product Details & Documents
These collapsible LDPE plastic containers have polypropylene closures and are convenient for space-saving and storage
Containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.
Cap size: 38-400.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • Level III Containers
    0.9 L (30.4 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $384.07
    $384.07
    Case of 12
       $384.07
    Supplier Number: EPC3030
  • Level III Containers
    3.8 L (1 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $290.07
    $290.07
    Case of 12
       $290.07
    Supplier Number: EPC3020
  • Level III Containers
    9.5 L (2.5 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 36
    Case of 36 - $980.31
    $980.31
    Case of 36
       $980.31
    Supplier Number: EPC3010
  • Level III Containers
    3.8 L (1 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 160
    Case of 160 - $1,825.37
    $1,825.37
    Case of 160
       $1,825.37
    Supplier Number: EPC3100
  • Level III Containers
    0.9 L (30.4 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 128
    Case of 128 - $2,475.81
    $2,475.81
    Case of 128
       $2,475.81
    Supplier Number: EPC3155