Cube® Inserts, Hedwin | Avantor
Cube® Inserts, Hedwin
Supplier:   Hedwin
Cube® Inserts, Hedwin
Supplier:  Hedwin
Certificates
Cube® Inserts, Hedwin
3 Options Available Below
Product Details & Documents
Durable, lightweight cube-shaped inserts are for use with Cubitainer® cartons (see 24300-655 series) for specimen collection and food or chemical storage. Inserts may also be used for stand-alone product packaging.
Top face accomodates 38mm (11/2") or 63mm (21/2") opening, and features molded-in reversible well that can be raised for pouring or depressed for storage
Amber color insert protects contents from UV rays and is ideal for patient collection of urine samples over 24-hour period. Features integrated handle, graduation marks, and labeling tag.
Cube® material (linear low density polyethylene blend) conforms to FDA Regulation 177.1520.
Non-glug design makes contents flow in continuous, uninterrupted stream. Containers collapse as product dispenses. Inserts are supplied with screw-on cap; natural color insert is supplied with glue-on neck.
Ordering information: Inserts are shipped folded into a space-saving nesting configuration. Inserts may be fluorine-treated to prevent chemical permeation, or "PR" coated for oxygen-sensitive food or chemical products; contact your Avantor sales representative for details.
Documents
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Color
    Availability
    Price
  • 0.9 L (0.2 gal.)
    With Screw Cap
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 128
    Case of 128 - $1,675.89
    $1,675.89
    Case of 128
       $1,675.89
    Supplier Number: CUB6050
  • 3.8 L (1 gal.)
    With Screw Cap
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 160
    Case of 160 - $5,065.73
    $5,065.73
    Case of 160
       $5,065.73
    Supplier Number: CUB6136
  • 3.8 L (1 gal.)
    Amber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $1,096.44
    $1,096.44
    Case of 100
       $1,096.44
    Supplier Number: CUB6057
Accessories
Informations sur le produit
  • Catalog #
    Weight
    Availability
    Price
  • Carton, Semi-White, Double Wall
    Carton, Semi-White, Double Wall
    56616-402
    275 lbs.
    Log in pour la disponibilité
    Pack of 1 - $32.32