Analyzer Sample Cups, Greiner Bio-One
Supplier: Greiner Bio-One
Jump to:
À propos de cet article
Sample cups are well suited for a number of established analyzer systems including Centrifichem, Coulter Counter®, Gemsaec, Hitachi®, Technicon™, and Sysmex® systems.
- Polyethylene or polystyrene construction
- Suited for a number of established analyzer systems
- Clear
Sample cup 82051-154 is constructed of polyethylene; all others are constructed of polystyrene.
2 Options Available Below
Product Details & Documents
Sample cups are well suited for a number of established analyzer systems including Centrifichem, Coulter Counter®, Gemsaec, Hitachi®, Technicon™, and Sysmex® systems.
- Polyethylene or polystyrene construction
- Suited for a number of established analyzer systems
- Clear
Sample cup 82051-154 is constructed of polyethylene; all others are constructed of polystyrene.
Product Family Options
Informations sur le produit
- DésignationCapacitéW×HAvailabilityPrice
- Sample Cup for Hitachi® Analyzers, Conical/Skirted1.5 ml (0.05 oz.)17×38 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 5,000Case of 5,000 - $298.74$298.74Prix promotionnel pour les nouveaux clientsCase of 5,000$298.74Supplier Number: 729101
- Sample Cup for Coulter Counter® Analyzers, Flat25 ml (0.85 oz.)32×60 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 1,250Case of 1,250 - $408.82$408.82Prix promotionnel pour les nouveaux clientsCase of 1,250$408.82Supplier Number: 668102