Barnstead Replacement Cartridges, Thermo Scientific
Barnstead Replacement Cartridges, Thermo Scientific
Barnstead Replacement Cartridges, Thermo Scientific
Barnstead Replacement Cartridges, Thermo Scientific
Barnstead Replacement Cartridges, Thermo Scientific
Cartridges feature all polypropylene construction and are designed to fit certain NANOpure®, E-pure™, B-pure™, PCS, Bantam holder, or cooling loop systems.
Product Family Options
  • Désignation
    Capacité
    For Use With
  • Cation Removal
    2500 Grains as NaCl
    B-Pure™, Cooling Loops, Bantam
    Each
    Each - $355.67
    $355.67
    Each
       $355.67
     : D0815
  • Organic Free Cartridge
    330 grains as NaCl
    NANOpure® Infinity
     : -
  • Ultrapure and Organic Removal with Mixed-Bed, Activated Carbon Resin
    750 Grains as NaCl
    B-Pure™, Still Pretreatment
    Each
    Each - $351.97
    $351.97
    Each
       $351.97
     : D0832
  • Organic Removal, Activated Carbon Resin
    5000 Liters Avg. Feed Water
    B-Pure™, Cooling Loops, Bantam
    Each
    Each - $463.13
    $463.13
    Each
       $463.13
     : D0813
  • Ultrapure, Mixed-Bed Resin
    760 grains as NaCl to a  1 Megohm Endpoint
    NANOpure® II, Older Style NANOpure® Systems, B-Pure™, Bantam, Cooling Loops
    Each
    Each - $622.16
    $622.16
    Each
       $622.16
     : D0809
  • MACROpure with Macroreticular, Activated Carbon Resin
    2000 gal.s
    NANOpure®, E-Pure™, Pretreatment for ORGANICfree
    Each
    Each - $576.28
    $576.28
    Each
       $576.28
     : D0836
  • ORGANICfree
    NANOpure® II, Older NANOpure® Systems
    Each
    Each - $564.82
    $564.82
    Each
       $564.82
     : D0820
  • Combination Colloid/Organic Removal with Macroreticular, Activated Carbon Resin
    5000 gal.s
    NANOpure®, E-Pure™, B-Pure™
    Each
    Each - $465.15
    $465.15
    Each
       $465.15
     : D0835
  • Pretreat Cartridge Tap+Di
    NANOpure® Infinity
     : -
  • Pretreat Cartridge R/O+Dist Fd
    NANOpure® Infinity
     : -
  • Oxygen Removal
    11 Liters S.T.P. (15 grams)
    B-Pure™, Cooling Loops
    Each
    Each - $448.58
    $448.58
    Each
       $448.58
     : D0811
  • Type1 Cartridge Kit Tap+Di Inf
    NANOpure® Infinity
     : -
  • Type1 Cartridge Kit R/O+Di Inf
    NANOpure® Infinity
     : -
  • ORGANICfree Cartridge Kit Tap+Diinf
    NANOpure® Infinity
     : -
  • ORGANICfree Cartridge Kit R/Odiinf
    NANOpure® Infinity
     : -
  • Pretreat Cartridge Kit
    NANOpure® Infinity
    Each
    Each - $3,024.02
    $3,024.02
    Each
       $3,024.02
     : D5026
  • High Capacity Cartridge, Two-Bed Ion Exchange Resin
    1760 grains as CaCO₃
    Bantam Deionizer Cartridge Systems, B-Pure™ Double/Single Holder and Harvey DI+ Cartridge Systems
    Each
    Each - $451.86
    $451.86
    Each
       $451.86
     : D0803