BD™ Universal Viral Transport System, BD Medical
CA90006-624
: BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:BD UVT 3 ml Vial
- Cat. no.:CA90006-624
The BD™ Universal Viral Transport System (UVT) is designed to transport viruses, chlamydiae, mycoplasmas, and ureaplasmas at room temperature. It is one of the only systems that may be stored and transported at 2 to 25 °C, all in one formulation.
- No refrigeration requirements: System does not require refrigerated storage or transport on ice, decreasing the risk of specimen rejection and eliminating messy specimen transport
- Reduced contamination: System features a unique blend of antimicrobics that target and suppress bacterial and fungal contamination without harming viruses, chlamydiae, mycoplasmas, or ureaplasmas
- Long-term storage: System helps store samples for long periods by preserving viruses and chlamydiae, when stored frozen with a cryoprotectant