VWR® Talon® Jumbo Holders
VWR® Talon® Jumbo Holders
VWR® Talon® Jumbo Holders
VWR® Talon® Jumbo Holders
Holder secures clamps to lab frames and ring stands and accepts rods up to 21mm (13/16") in diameter.
  • Electropolished stainless steel or aluminum construction
  • Secure support of clamps
  • Accepts rods of a range of diameters
Jumbo holders provide sturdy attachments of clamps to lab frames and ring stands.
Product Family Options
  • Désignation
    Matière
  • Clamp Holder
    Stainless steel
    Each
    Each - $107.26
    $107.26
    Each
       $107.26
  • Clamp Holder
    Aluminum
    Each
    Each - $72.90
    $72.90
    Case of 25
    Case of 25 - $1,822.60
    $1,822.60
    Each
       $72.90
    Case of 25
       $1,822.60