BD Falcon™ SWUBE™ Collection and Transport System, BD Diagnostics
: BD
Combination swabs and tubes provide a simple and effective method of collection, transporting, protecting, and storing specimens
Sterile, individually wrapped tubes are provided without media, but can be used with a wide array of reagents, media, or solutions. The 14.6–15cm (53/4–6”) long shafts reach touch sampling sites. Available with screw caps or friction caps
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together