Super Erecta® Stem Casters, Metro™
Supplier:   Metropolitan Wire
Super Erecta® Stem Casters, Metro™
Supplier:  Metropolitan Wire
Certificates
Super Erecta® Stem Casters, Metro™
Super Erecta® Stem Casters, Metro™
22 Options Available Below
Product Details & Documents
For use with Super Erecta® shelving and SiteSelect® posts to create mobile units. Swivel, foot-operated brake, and rigid styles are available.
  • Resilient rubber tread casters are non-marking and feature a molded, soft tread that provides quiet operation
  • Polyurethane tread casters are abrasion-resistant, non-marking, and shock-absorbant
Casters with high modulus rubber tread provide extra shock protection and have excellent resistance to oil and grease. Special cart-washable casters are constructed of stainless steel components. Casters include grease seals and zerk fittings to withstand high-pressure washings.
Conductive casters conform to Federal Specification FF-C-88C regarding the acceptable standards for conductive wheels. To ensure proper conductivity, aluminum split sleeves must be used with each shelf.
 : Rigid casters are held in position by a connecting channel; when ordering rigid casters, shelf width must be provided. An 8.9 cm (3¹/₂") diameter donut is furnished standard on each caster.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Matière
    Diameter
    Diameter × Tread Width
    Largeur
    Hauteur
    Load Capacity
    Weight
    Shipping Weight
    Availability
    Price
  • Rigid stem caster, resilient rubber
    Silver/black
    Heavy gauge cold rolled steel zinc plated horn; resilient rubber tread
    5"
    200 lbs.
    2 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $144.18
    $144.18
    Each
       $144.18
    Supplier Number: 5MR
  • Rigid stem caster, high modulus rubber donut tread
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn. Advantage high modulus gray thermoplastic rubber donut tread; polyolefin hub
    5"
    250 lbs.
    2.4 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $159.55
    $159.55
    Each
       $159.55
    Supplier Number: 5MDRA
  • Rigid stem caster, high modulus rubber donut tread
    Silver/grey
    Stainless steel horn; Advantage hi-modulus gray thermoplastic rubber donut tread; polyolefin hub
    5"
    250 lbs.
    2.4 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $574.00
    $574.00
    Each
       $574.00
    Supplier Number: 5MDRGSA
  • Rigid stem caster, polyurethane
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; polyurethane flat tread; polyethylene hub
    5"
    300 lbs.
    2 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $207.20
    $207.20
    Each
       $207.20
    Supplier Number: 5MPR
  • Rigid stem caster, polyurethane
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; polyurethane flat tread; polyethylene hub
    6"
    400 lbs.
    2.3 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.93
    $290.93
    Each
       $290.93
    Supplier Number: 6MPR
  • Swivel stem caster, high modulus rubber donut tread
    Silver/grey
    Stainless steel horn; Advantage hi-modulus gray thermoplastic rubber donut tread; polyolefin hub
    5"
    250 lbs.
    2.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $545.79
    $545.79
    Each
       $545.79
    Supplier Number: 5MDGSA
  • Swivel stem caster, polyurethane
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; polyurethane flat tread; polyethylene hub
    5"
    300 lbs.
    2.1 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $185.53
    $185.53
    Each
       $185.53
    Supplier Number: 5MP
  • Swivel stem caster, polyurethane
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; polyurethane flat tread; polyethylene hub
    6"
    400 lbs.
    2.3 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $248.42
    $248.42
    Each
       $248.42
    Supplier Number: 6MP
  • Swivel stem caster, resilient rubber
    Silver/black
    Heavy gauge cold rolled steel nickel plated horn; resilient rubber tread
    4"
    125 lbs.
    2 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $91.33
    $91.33
    Each
       $91.33
    Supplier Number: 4LD
  • Swivel stem caster, resilient rubber
    Silver/black
    Heavy gauge cold rolled steel nickel plated horn; resilient rubber tread
    5"
    125 lbs.
    2 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.01
    $103.01
    Each
       $103.01
    Supplier Number: 5LD
  • Swivel stem caster, resilient rubber
    Silver/black
    Heavy gauge cold rolled steel zinc plated horn. resilient rubber tread
    5"
    200 lbs.
    2.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.72
    $119.72
    Each
       $119.72
    Supplier Number: 5M
  • Swivel/brake stem caster, high modulus rubber donut tread
    Silver/grey
    Stainless steel horn; Advantage hi-modulus gray thermoplastic rubber donut tread; polyolefin hub
    5"
    250 lbs.
    2.6 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $574.00
    $574.00
    Each
       $574.00
    Supplier Number: 5MDBGSA
  • Swivel/brake stem caster, polyurethane
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; polyurethane flat tread; polyethylene hub
    6"
    400 lbs.
    2.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.93
    $290.93
    Each
       $290.93
    Supplier Number: 6MPB
  • Swivel/brake stem caster, resilient rubber
    Silver/black
    Heavy gauge cold rolled steel zinc plated horn. resilient rubber tread
    5"
    200 lbs.
    2.6 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.40
    $142.40
    Each
       $142.40
    Supplier Number: 5MB
  • Brake caster, conductive wheel tread
    5"
    5×1¼"
    175 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $212.27
    $212.27
    Each
       $212.27
    Supplier Number: 5MFA
  • Brake caster, conductive wheel tread
    5"
    5×1¼"
    200 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $233.84
    $233.84
    Each
       $233.84
    Supplier Number: 5MFBA
  • Swivel stem caster, high modulus rubber donut tread
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; Advantage high modulus gray thermoplastic rubber donut tread; Polyolefin hub
    5"
    250 lbs.
    2.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $137.64
    $137.64
    Each
       $137.64
    Supplier Number: 5MDA
  • Brake caster, conductive wheel tread
    5"
    5×1¼"
    200 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $179.45
    $179.45
    Each
       $179.45
    Supplier Number: 5MESD
  • Chrome-Plated Mobile Post
    1"
    27"
    0.75 kg (1.75 lbs.)
    2.1 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $195.73
    $195.73
    Each
       $195.73
    Supplier Number: 5MBESD
  • Chrome-Plated Mobile Post
    1"
    33"
    0.9 kg (2 lbs.)
    2.1 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $205.93
    $205.93
    Each
       $205.93
    Supplier Number: 5MC
  • Chrome-Plated Mobile Post
    1"
    63"
    1.6 kg (3.5 lbs.)
    3.5 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $231.76
    $231.76
    Each
       $231.76
    Supplier Number: 5MBC
  • Swivel/brake stem caster, high modulus rubber donut tread
    Silver/grey
    Heavy gauge cold rolled steel nickel plated horn; Advantage high modulus gray thermoplastic rubber donut tread; polyolefin hub
    5"
    250 lbs.
    2.6 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $159.49
    $159.49
    Each
       $159.49
    Supplier Number: 5MDBA