SmartWall Plus™ Storage System, Metro™ | Avantor
SmartWall Plus™ Storage System, Metro™
Chrome
Catalog # CA82023-314
Supplier:   Metropolitan Wire
Fournisseur #:  1230CSNC
SmartWall Plus™ Storage System, Metro™
Catalog # CA82023-314
Supplier:  Metropolitan Wire
SmartWall Plus™ Storage System, Metro™
SmartWall Plus™ Storage System, Metro™
Chrome
Price
$226.47
Log in pour la disponibilité
Each
Product Details & Documents
Designed to optimize wall storage through a variety of shelf, accessory grid, and cabinet combinations organized onto easy-to-install wall tracks.
  • Corrosion-proof Type 304 stainless steel components
  • Cantilever shelves attach directly to posts without shelf supports
  • Taupe post assemblies including steel brackets are coated in corrosion-resistant taupe epoxy
Corrosion-resistant taupe epoxy components address corrosion and durability issues and in a multitude of applications. Wall tracks are available in 16 gauge Type 304 stainless steel and corrosion-resistant taupe epoxy finishes. Holes on wall tracks are spaced 20.3cm (8") apart; first and last holes are positioned 10.2cm (4") from track ends. Weight capacities per unit depend on the method of mounting and wall structure. Joining braces must be used to connect adjacent tracks. Post assemblies must be secured to wall surfaces through the bottom bracket. Smartwall Plus™ is only to be used with shelves that are 45.7cm (18") deep or less. Brackets for chrome, Metroseal 3, and stainless steel post assemblies are Type 304 stainless steel.
Ordering information: Wall tracks are supplied with two aluminum joining braces/stops, two self-tapping stainless steel screws for joining braces, and two plastic end caps. Post assemblies include top bracket, intermediate bracket, bottom bracket, standard Super Erecta® post, and stainless steel bolt for bottom bracket. For additional SmartWall Plus™ storage and accessory items, see 83009-610 series. For easy-order starter kits, see 82023-520 series. Super Erecta® shelves require shelf supports, two single supports per shelf. Double shelf supports are intended for a run of multiple shelves. Super Erecta® shelves are available in additional finishes (see 19766-070, 19766-072, and 83009-332 series). MetroMax™ corner adapters are required to allow MetroMax™ shelves to be used with standard shelf supports.
Spécifications
  • Finish:
    Chrome
Accessories
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal™ 3 Slanted Lid Holder/Drying Shelf
    Metroseal™ 3 Slanted Lid Holder/Drying Shelf
    CA82023-640
    5 lbs.
    Log in pour la disponibilité
    Each - $261.05
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Plated Flatware/Instrument Cylinder
    Plated Flatware/Instrument Cylinder
    CA82023-642
    0.3 lbs.
    Log in pour la disponibilité
    Each - $38.80
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Plated Cylinder Holder
    Plated Cylinder Holder
    CA82023-644
    1 lbs.
    Log in pour la disponibilité
    Each - $85.42
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-610
    12 lbs.
    Log in pour la disponibilité
    Each - $177.62
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-612
    14 lbs.
    Log in pour la disponibilité
    Each - $220.99
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-614
    12 lbs.
    Log in pour la disponibilité
    Each - $246.90
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-616
    15.5 lbs.
    Log in pour la disponibilité
    Each - $304.88
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Shelf with Retaining Edge for Grid
    Metroseal 3 Shelf with Retaining Edge for Grid
    CA82023-622
    12 lbs.
    Log in pour la disponibilité
    Each - $311.46
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Shelf with Retaining Edge for Grid
    Metroseal 3 Shelf with Retaining Edge for Grid
    CA82023-624
    16 lbs.
    Log in pour la disponibilité
    Each - $378.11
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Shelf with Retaining Edge for Grid
    Metroseal 3 Shelf with Retaining Edge for Grid
    CA82023-626
    15 lbs.
    Log in pour la disponibilité
    Each - $311.46
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Shelf with Retaining Edge for Grid
    Metroseal 3 Shelf with Retaining Edge for Grid
    CA82023-628
    18 lbs.
    Log in pour la disponibilité
    Each - $350.44
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Shelf with Retaining Edge for Grid
    Metroseal 3 Shelf with Retaining Edge for Grid
    CA82023-630
    24 lbs.
    Log in pour la disponibilité
    Each - $418.47
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Flat Grid Shelf for Grid
    Metroseal 3 Flat Grid Shelf for Grid
    CA82023-632
    7 lbs.
    Log in pour la disponibilité
    Each - $287.61
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Pronged Hook
    Metroseal 3 Pronged Hook
    CA82023-634
    0.3 lbs.
    Log in pour la disponibilité
    Each - $40.09
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Bottle Holder, 6-Prong
    Metroseal 3 Bottle Holder, 6-Prong
    CA82023-636
    2.3 lbs.
    Log in pour la disponibilité
    Each - $136.64
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal™ 3 Tray Drying Rack
    Metroseal™ 3 Tray Drying Rack
    CA82023-638
    12 lbs.
    Log in pour la disponibilité
    Each - $487.23
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-608
    9 lbs.
    Log in pour la disponibilité
    Each - $143.11
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-618
    12.8 lbs.
    Log in pour la disponibilité
    Each - $248.29
Informations sur le produit
  • Catalog #
    Shipping Weight
    Availability
    Price
  • Metroseal 3 Grid
    Metroseal 3 Grid
    CA82023-620
    16.5 lbs.
    Log in pour la disponibilité
    Each - $305.47